EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Dual-responsive supramolecular hydrogels from water-soluble PEG-grafted copolymers and cyclodextrin.
PMID 19544291
Abstract
Novel temperature and pH dual-responsive hydrogels were constructed by inclusion of poly(PEGMA)-co-poly(DMA) with alpha-cyclodextrin in aqueous solution. The temperature- or pH-induced sol/gel transition in the hydrogels was completely reversible. Studies on structure/property relationships show that chain uniformity, graft density and copolymer concentration affect the hydrogel behavior. A dual-responsive mechanism is proposed. The in vitro release of a model drug from this hydrogel was studied. It was found that the release kinetics were greatly accelerated at higher temperature and at acidic pH conditions, indicating potential applications in controlled drug delivery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0531
5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein hydrochloride, ≥90% (HPLC)
C23H12Cl2N4O5 · HCl