EMAIL THIS PAGE TO A FRIEND
Chemical research in toxicology
Preparation and properties of lithium and sodium peroxynitrite.
PMID 19548349
Abstract
We describe the preparation of aqueous solutions of LiONOO and NaONOO from (Me4N)ONOO. An aqueous solution of analytically pure, commercially available (Me4N)ONOO is applied to an Amberlyst 15 column at 4 degrees C, and the Me4N+ is rapidly (in 20 min) exchanged against Li+ or Na+. The exchanged peroxynitrite is produced in yields of ca. 90% with a nitrite content of 5-6%.