EMAIL THIS PAGE TO A FRIEND
Breast cancer research : BCR
Expression of FOXA1 and GATA-3 in breast cancer: the prognostic significance in hormone receptor-negative tumours.
PMID 19549328
Abstract
The expression of additional genes, other than oestrogen receptor (ER), may be important to the hormone-responsive phenotype of breast cancer. Microarray analyses have revealed that forkhead box A1 (FOXA1) and GATA binding protein 3 (GATA-3) are expressed in close association with ERalpha, both encoding for transcription factors with a potential involvement in the ERalpha-mediated action in breast cancer. The purpose of this study was to explore if the expression of FOXA1 and GATA-3 may provide an opportunity to stratify subsets of patients that could have better outcome, among the ERalpha-negative/poor prognosis breast cancer group. We evaluate FOXA1 and GATA-3 expression in 249 breast carcinomas by immunohistochemistry, associating it with breast cancer molecular markers, clinicopathological features and patient's survival. The clinicopathological features and immunohistochemical markers of the tumours were compared using the chi-square test and ANOVA. Disease-free survival was analysed through Kaplan-Meier survival curves and Cox regression. FOXA1 expression was demonstrated in 42% of invasive carcinomas, while GATA-3 was detected in 48% of the cases. FOXA1 expression was inversely associated with tumour size, Nottingham Prognostic Index, histological grade, lymph vascular invasion, lymph node stage and human epidermal growth factor receptor-2 (HER-2) overexpression, while GATA-3 expression showed inverse association with histological grade and HER-2. Both FOXA1 and GATA-3 were directly associated with ERalpha and progesterone receptor. Among FOXA1-positive tumours, 83.1% are comprised in the luminal A subtype, similar to GATA-3 where 87.7% of positive tumours were classified within this molecular subtype. In the subset of ERalpha-negative patients, those who were FOXA1-negative had a 3.61-fold increased risk of breast cancer recurrence when compared with the FOXA1-positive. FOXA1 was a significant predictor of good outcome in breast cancer, whereas GATA-3 was an important luminal marker. The expression of FOXA1 may be used for risk stratification among ERalpha-negative patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
405M-1
FOXA1 (2F83) Mouse Monoclonal Antibody