EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Species differences for stereoselective metabolism of ethofumesate and its enantiomers in vitro.
PMID 19552529
Abstract
1. The stereoselective metabolism of ethofumesate (ETO) and its enantiomers in rabbit and rat liver microsomes have been studied by chiral high-performance liquid chromatography (HPLC) method. Two metabolites were detected in both liver microsomes in the presence of beta-nicotinamide adenine dinucleotide phosphate (NADPH). 2. The T(1/2) of (+)-ETO and (-)-ETO in rabbit liver microsomes were 12.2 and 4.7 min of rac-ETO and 25.9 and 6.7 of ETO enantiomers. However, the T(1/2) of (+)-ETO and (-)-ETO in rat liver microsomes were 5.3 and 5.9 min of rac-ETO and 7.8 and 10.6 of ETO enantiomers. The stereoselective selectivity is similar to the in vivo study. 3. After incubation of ETO enantiomers, stereoselectivity was present in the formation of ETO-OH enantiomer in rabbit liver microsomes, but stereoselectivity was not evident in rat liver microsomes. 4. There was no chiral inversion from the (+)-ETO to (-)-ETO or inversion from (-)-ETO to (+)-ETO in both rabbit and rat liver microsomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45479
Ethofumesate, PESTANAL®, analytical standard
C13H18O5S