EMAIL THIS PAGE TO A FRIEND
Clinical interventions in aging
Six-month depot formulation of leuprorelin acetate in the treatment of prostate cancer.
PMID 19554097
Abstract
Hormonal deprivation therapy is well established for the treatment of locally advanced and metastatic prostate cancer, as well as the adjuvant treatment of some patients with localized disease. Long-acting gonadotropin releasing hormone (GnRH) agonists have become a mainstay of androgen deprivation therapy, due to their efficacy, tolerability, and convenience of use. One-month, 3-month, and 4-month depot leuprorelin formulations are well established and widely used to this end. Recently, a 6-month depot leuprorelin has been approved for use in advanced and metastatic prostate cancer patients. With similar efficacy and side effect profiles to earlier formulations, 6-month depot leuprorelin is a convenient treatment option for these patients. This review will highlight the role of GnRH agonists in the treatment of prostate cancer with a focus on the clinical efficacy, pharmacology, and patient-focused outcomes of the newer 6-month 45 mg depot leuprorelin formulation in comparison to available shorter-acting products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L0376000 Leuprorelin, European Pharmacopoeia (EP) Reference Standard
C59H84N16O12