EMAIL THIS PAGE TO A FRIEND
Tetrahedron letters
Enantioselective Reduction of Prochiral Ketones using Spiroborate Esters as Catalysts.
PMID 19554205
Abstract
Novel spiroborate esters derived nonracemic 1,2-aminoalcohols and ethylene glycol are reported as highly effective catalysts for the asymmetric borane reduction of a variety of prochiral ketones with borane-dimethyl sulfide complex at room temperature. Optically active alcohols were obtained in excellent chemical yields using 0.1 to 10 mol % of catalysts with up to 99% ee.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
192120
Borane dimethyl sulfide complex solution, 2.0 M in THF
C2H9BS