EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Tetrahydrocarbazole amides with potent activity against human papillomaviruses.
PMID 19556128
Abstract
Synthesis of a series of tetrahydrocarbazole amides with potent activity against human papillomaviruses is described. Synthetic approaches allowing for variation of the substitution pattern of the tetrahydrocarbazole and the amide are outlined and resulting changes in antiviral activity and certain developability parameters are highlighted. Several compounds with in vitro antiviral activity (W12 antiviral assay) in the single digit nanomolar range were identified and N-[(1R)-6-chloro-2,3,4,9-tetrahydro-1H-carbazol-1-yl]-2-pyridinecarboxamide was selected for further evaluation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1824
GSK983, ≥98% (HPLC)
C18H16ClN3O
SML1825
GSK984, ≥98% (HPLC)
C18H16ClN3O