EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Fully automated method for simultaneous determination of total cysteine, cysteinylglycine, glutathione and homocysteine in plasma by HPLC with UV absorbance detection.
PMID 19559659
Abstract
A fully automated HPLC method for the simultaneous determination of total thiols in plasma samples has been developed. The method involves reductive conversion of disulfides to their reduced counterparts with the use of tris(2-carboxyethyl)phosphine. After reduction the newly formed sulfhydryl groups are reacted with 2-chloro-1-methylquinolinium tetrafluoroborate to form 2-S-quinolinium derivatives followed by deproteinization by dialysis. The reaction products are separated by reversed-phase HPLC, detected and quantified by UV absorbance detection at 355nm. The recommended HPLC procedure enables measurement of four main plasma aminothiols cysteine, cysteinylglycine, glutathione, and homocysteine with low imprecision (mean relative standard deviations within calibration range, 3.47%, 5.34%, 4.25% and 3.26%, respectively) and good sensitivity. Accuracy, expressed as the mean measured amount as percentage of added amount, was within 97.5-103.0%, 98.3-102.5%, 96.3-99.5% and 97.1-99.1%, respectively. The lower limit of quantification for all thiols was 0.5microM. The whole unattended instrument acquisition time amounts 13min.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0166
Cys-Gly, ≥85% (TLC)
C5H10N2O3S