EMAIL THIS PAGE TO A FRIEND
Talanta
Comments on "Determination of S-nitrosoglutathione and other nitrosothiols by p-hydroxymercurybenzoate derivatization and reverse phase chromatography coupled with chemical vapor generation atomic fluorescence detection" by Bramanti et al.
PMID 19559920
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0166
Cys-Gly, ≥85% (TLC)
C5H10N2O3S