EMAIL THIS PAGE TO A FRIEND
Clinical and experimental pharmacology & physiology
Adenosine receptor interactions alter cardiac contractility in rat heart.
PMID 19566843
Abstract
1. The effect of the adenosine A(2) receptor (AdoA(2)R) agonist N(6)-[2-(3,5-dimethoxyphenyl)-2-(2-methylphenyl)-ethyl]adenosine (DPMA) on adenosine A(1) receptor (AdoA(1)R)-mediated negative inotropic responses was investigated in rat heart. 2. Hearts from male Wistar rats (250-350 g) were perfused with Krebs'-Henseleit solution at constant flow in non-recirculating Langendorff mode. Hearts were paced at 5 Hz (5 ms duration, supramaximal voltage) via ventricular electrodes. After 30 min equilibration, (R)-N(6)-phenylisopropyl adenosine (R-PIA) concentration-response curves were constructed in the absence or presence of DPMA. 3. In paced hearts, R-PIA induced concentration-dependent decreases in triple product (heart rate x peak systolic developed pressure x dP / dt(max)), which were significantly attenuated by 1 nmol / L DPMA with a shift in pEC(50) from 8.0 +/- 0.5 (n = 9) in control hearts to 6.63 +/- 1.03 (n = 5) in treated tissues (P < 0.05). The AdoA(2A)R antagonist 8-(3-chlorostyryl)caffeine (1 micromol / L) and the adenylyl cyclase inhibitor cis-N-(2-phenylcyclopentyl)-azacyclotridec-1-en-2-amine hydrochloride (MDL12330A; 100 nmol / L) reversed the effects of DPMA on AdoA(1)R-mediated negative inotropic actions, whereas the AdoA(2B)R antagonist alloxazine (3 micromol / L) had no effect on DPMA activity. 4. The results of the present study show that stimulation of the AdoA(2)R attenuates AdoA(1)R-dependent reductions in inotropic state. The receptor involved appears to be the AdoA(2A)R and its action involves stimulation of adenylyl cyclase activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A28651
Alloxazine, 96%
C10H6N4O2
M182
MDL-12,330A hydrochloride, ≥98% (HPLC), powder
C23H36N2 · HCl