EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
A modified four-state model for the "dual fluorescence" of N(6),N(6)-dimethyladenine derived from femtosecond fluorescence spectroscopy.
PMID 19569655
Abstract
The radiationless deactivation of the excited electronic states of the dual fluorescence molecule N(6),N(6)-dimethyladenine (DMAde) was investigated using femtosecond time-resolved fluorescence up-conversion spectroscopy. The molecules were studied in solution in water and in dioxane. Fluorescence-time profiles were recorded in the wide wavelength range of 290
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2629
6-(Dimethylamino)purine, ≥98%
C7H9N5