EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
1-alkyl-8-(piperazine-1-sulfonyl)phenylxanthines: development and characterization of adenosine A2B receptor antagonists and a new radioligand with subnanomolar affinity and subtype specificity.
PMID 19569717
Abstract
A new series of 1-alkyl-8-(piperazine-1-sulfonyl)phenylxanthines was designed, synthesized, and characterized in radioligand binding and functional assays at A(2B) adenosine receptors. A(2B) antagonists with subnanomolar affinity and high selectivity were discovered. The most potent compounds were 1-ethyl-8-(4-(4-(4-trifluoromethylbenzyl)piperazine-1-sulfonyl)phenyl)xanthine (24, PSB-09120, K(i) (human A(2B)) = 0.157 nM) and 8-(4-(4-(4-chlorobenzyl)piperazine-1-sulfonyl)phenyl)-1-propylxanthine (17, PSB-0788, K(i) (human A(2B)) = 0.393 nM). Moreover, 8-(4-(4-(4-chlorophenyl)piperazine-1-sulfonyl)phenyl)-1-propylxanthine (35, PSB-603) was developed as an A(2B)-specific antagonist exhibiting a K(i) value of 0.553 nM at the human A(2B) receptor and virtually no affinity for the human and rat A(1) and A(2A) and the human A(3) receptors up to a concentration of 10 microM. A tritiated form of the compound was prepared as a new radioligand and characterized in kinetic, saturation, and competition studies. It was shown to be a useful pharmacological tool for the selective labeling of human as well as rodent A(2B) receptors (K(D) human A(2B) 0.403 nM, mouse A(2B) 0.351 nM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5134
2-Chloroadenosine
C10H12ClN5O4
27602
Caffeine, anhydrous, tested according to Ph.Eur.
C8H10N4O2
SML1983 PSB-603, ≥98% (HPLC)
C24H25ClN6O4S