EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A simple and convenient copper-catalyzed tandem synthesis of quinoline-2-carboxylates at room temperature.
PMID 19572501
Abstract
We developed a simple and convenient copper-catalyzed method for the synthesis of quinoline-2-carboxylate derivatives through sequential intermolecular addition of alkynes onto imines and subsequent intramolecular ring closure by arylation. The efficiency of this system allowed the reactions to be carried out at room temperature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
174823
4-Quinolinecarboxylic acid, 97%
C10H7NO2