EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis and spectroscopic studies of the charge transfer complexes of 2- and 3-aminopyridine.
PMID 19574089
Abstract
The interactions of the electron donors 2-aminopyridine (2APY) and 3-aminopyridine (3APY) with the pi-acceptors tetracyanoethylene (TCNE), 2,3-dichloro-5,6-dicyano-p-benzoquinone (DDQ), 2-chloro-1,3,5-trinitrobenzene (picryl chloride, PC), and 2,3,5,6-tetrachloro-1,4-benzoquinone (chloranil) were studied spectrophotometrically in chloroform at room temperature. The electronic and infrared spectra of the formed molecular charge transfer (CT) complexes were recorded. Photometric titration showed that the stoichiometries of the reactions were fixed and depended on the nature of both the donor and the acceptor. The molecular structures of the CT-complexes were, however, independent of the position of the amino group on the pyridine ring and were formulated as [(APY)(TCNE)], [(APY)(DDQ)], [(APY)(PC)], and [(APY) (chloranil)]. The formation constants (K(CT)), charge transfer energy (E(CT)) and molar extinction coefficients (epsilon(CT)) of the formed CT-complexes were obtained.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A78209
3-Aminopyridine, 99%
C5H6N2
T8809
Tetracyanoethylene, 96%
C6N4