EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Analysis of vibrational spectra of 2 and 3-methylpiperidine based on density functional theory calculations.
PMID 19574090
Abstract
The experimental and theoretical vibrational spectra of 2 and 3-methylpiperidine (abbreviated as 2-MP and 3-MP) were studied. The FT-Infrared spectra of 2-MP and 3-MP molecules were recorded in the liquid phase. The structural and spectroscopic analysis of the title molecules were made by using density functional harmonic calculations. For the title molecules, only one form was found most stable structure by using B3LYP level with the 6-311G (d,p) basis set. Selected experimental bands were assigned and characterized based on the scaled theoretical wave numbers by their total energy distribution (TED).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M72803
2-Methylpiperidine, 98%
C6H13N
W424401
2-Methylpiperidine, 98%
C6H13N