EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Fe3O4 core/layered double hydroxide shell nanocomposite: versatile magnetic matrix for anionic functional materials.
PMID 19575433
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
400866
Iron(II) oxide, −10 mesh, 99.7% trace metals basis
FeO