EMAIL THIS PAGE TO A FRIEND
Journal of Virological Methods
Evaluation of the NucliSens EasyQ HIV-1 v1.1 and RealTime HIV-1 kits for quantitation of HIV-1 RNA in plasma.
PMID 19576640
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
3110 LIGHT DIAGNOSTICS Respiratory Virus Panel I DFA, ~50 tests