EMAIL THIS PAGE TO A FRIEND
Annals of tropical medicine and parasitology
Successful use of miltefosine and sodium stibogluconate, in combination, for the treatment of an HIV-positive patient with visceral leishmaniasis: a case report and brief review of the literature.
PMID 19583915