EMAIL THIS PAGE TO A FRIEND
Journal of cell science
AIMP2 promotes TNFalpha-dependent apoptosis via ubiquitin-mediated degradation of TRAF2.
PMID 19584093
Abstract
AIMP2 (aminoacyl-tRNA synthetase interacting multifunctional protein 2; also known as JTV-1) was first identified as p38 in a macromolecular protein complex that consisted of nine different aminoacyl-tRNA synthetases and two other auxiliary factors. AIMP2 also plays pivotal roles in the regulation of cell proliferation and death. Although AIMP2 was previously shown to augment TNFalpha-induced cell death, its working mechanism in this signal pathway was not understood. Here, we investigate the functional significance and mode of action of AIMP2 in TNFalpha signaling. TNFalpha-induced cell death was compromised in AIMP2-deficient or -suppressed cells and exogenous supplementation of AIMP2 augmented apoptotic sensitivity to TNFalpha signaling. This activity was confirmed by the AIMP2-dependent increase of IkappaB and suppression of NFkappaB. We found binding of AIMP2 to TRAF2, a key player in the TNFalpha signaling pathway. AIMP2 augmented the association of an E3 ubiquitin ligase, c-IAP1, with TRAF2, causing ubiquitin-dependent degradation of TRAF2. These findings suggest that AIMP2 can mediate the pro-apoptotic activity of TNFalpha via the downregulation of TRAF2 expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3501052
Anti-AIMP2 antibody produced in rabbit, affinity isolated antibody