EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Polyionic polymers--heterogeneous media for metal nanoparticles as catalyst in Suzuki-Miyaura and Heck-Mizoroki reactions under flow conditions.
PMID 19590749
Abstract
The preparation of monolithic polyionic supports which serve as efficient heterogeneous supports for palladium(0) nanoparticles is described. These functionalized polymers were incorporated inside a flow reactor and employed in Suzuki-Miyaura and Heck cross couplings under continuous flow conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357804
4′-Iodoacetophenone, ≥97%
C8H7IO