EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Dihydropyridine C-glycoconjugates by organocatalytic Hantzsch cyclocondensation. Stereoselective synthesis of alpha-threofuranose C-nucleoside enantiomers.
PMID 19590795
Abstract
The Hantzsch reaction of C-glycosyl aldehyde/enamino ester/beta-ketoester systems under l-proline catalysis to give dihydropyridine C-glycoconjugates is reported. Asymmetric cyclocondensations of differentially substituted enamine and beta-dicarbonyl components with formyl alpha-L-C-threofuranoside and with the alpha-D-isomer were also carried out. Each reaction occurred with high yet opposite stereoselectivity (de >95%) so that the pair of alpha-threofuranose C-nucleoside enantiomers was prepared.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7392
D-(−)-Threose, ≥60%, syrup
C4H8O4
T7642
L-(+)-Threose, ≥60%, syrup
C4H8O4