EMAIL THIS PAGE TO A FRIEND
Molecular medicine (Cambridge, Mass.)
The selective alpha7 agonist GTS-21 attenuates cytokine production in human whole blood and human monocytes activated by ligands for TLR2, TLR3, TLR4, TLR9, and RAGE.
PMID 19593403
Abstract
The cholinergic antiinflammatory pathway modulates inflammatory cytokine production through a mechanism dependent on the vagus nerve and the alpha7 subunit of the nicotinic acetylcholine receptor. GTS-21 [3-(2,4-dimethoxybenzylidene) anabaseine], a selective alpha7 agonist, inhibits inflammatory cytokine production in murine and human macrophages and in several models of inflammatory disease in vivo, but to date its antiinflammatory efficacy in human monocytes has not been characterized. We report here our findings that GTS-21 attenuates tumor necrosis factor (TNF) and interleukin 1beta levels in human whole blood activated by exposure to endotoxin. GTS-21 inhibited TNF production in endotoxin-stimulated primary human monocytes in vitro at the transcriptional level. The suppressive effect of GTS-21 was more potent than nicotine in whole blood and monocytes. Furthermore, GTS-21 attenuated TNF production in monocytes stimulated with peptidoglycan, polyinosinic-polycytidylic acid, CpG, HMGB1 (high-mobility group box 1 protein), and advanced glycation end product-modified albumin. GTS-21 decreased TNF levels in endotoxin-stimulated whole blood obtained from patients with severe sepsis. These findings establish the immunoregulatory effect of GTS-21 on human monocytes, and indicate the potential benefits of further exploration of GTS-21's therapeutic uses in human inflammatory disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0326
GTS-21, ≥97% (HPLC)
C19H20N2O2 · 2HCl