EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Design, synthesis, and bioactivity study of novel benzoylpyridazyl ureas.
PMID 19601668
Abstract
A series of novel benzoylpyridazyl ureas were designed and synthesized from maleic anhydride and hydrazine monohydrate. These benzoylureas were identified by (1)H NMR spectroscopy and element analysis. The bioactivities of the new compounds were evaluated. These compounds exhibited larvicidal activities against oriental armyworm, and in particular, compound 13 displayed comparable activity to the commercial insecticide Hexaflumuron. Most of these compounds also had some larvicidal activities against mosquito. Interestingly, some compounds showed good plant growth regulatory activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37902
Hexaflumuron, PESTANAL®, analytical standard
C16H8Cl2F6N2O3