EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A general copper-catalyzed coupling of azoles with vinyl bromides.
PMID 19603753
Abstract
A copper-catalyzed methodology for the coupling of vinyl bromides with azoles has been developed. This protocol uses a combination of 10 mol % of copper iodide and 20 mol % of ethylenediamine as catalyst. The reaction proceeds with various azoles and vinyl bromides, and the double bond geometry of vinyl bromides is retained under the reaction conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1902
Vinyl bromide, 98%
C2H3Br
434191
Vinyl bromide solution, 1.0 M in THF
C2H3Br