EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
1,4-Dioxane-2,5-dione-type monomers derived from l-ascorbic and d-isoascorbic acids. Synthesis and polymerisation.
PMID 19604503
Abstract
l-Ascorbic and d-isoascorbic acids have been used as the starting materials for the preparation of (3R,4'S)-3-(2',2'-dimethyl-1',3'-dioxolan-4'-yl)-1,4-dioxane-2,5-dione (IPTA), (3R and S, 4'S,6R)-3-methyl-6-(2',2'-dimethyl-1',3'-dioxolan-4'-yl)-1,4-dioxane-2,5-dione (IPTP) and (3R,4'R)-3-(2',2'-dimethyl-1',3'-dioxolan-4'-yl)-1,4-dioxane-2,5-dione (IPEA), three novel 1,4-dioxane-2,5-dione-type monomers. Ring-opening homopolymerisation and copolymerisation of the IPTA monomer, derived from l-ascorbic acid, with d,l-lactide have been performed. The polymers were characterised by elemental microanalysis, as well as IR and (1)H and (13)C NMR spectroscopies. GPC was used to estimate product molecular weights, and thermal studies (DSC and TGA) revealed that all the polymers were amorphous, being stable up to 250 degrees C under nitrogen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W241008
D-Isoascorbic acid, FCC, FG
C6H8O6
856061
D-(−)-Isoascorbic acid, 98%
C6H8O6