EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological sciences
Decrease in the kainate-induced wet dog shake behavior in genetically epilepsy-prone rats: possible involvement of an impaired synaptic transmission to the 5-HT(2A) receptor.
PMID 19609070
Abstract
Genetically epilepsy-prone rats (GEPR-9s) were derived from Sprague-Dawley rats (SD). The number of kainate-induced wet dog shake behavior (WDS) responses was found to decrease significantly in GEPR-9s compared to SD. WDS responses were potentiated by 5-hydroxytryptophan or 2,5-dimethoxy-4-iodoamphetamine and antagonized by ritanserin. The antagonizing effect of ritanserin on WDS latency was more evident in GEPR-9s than in SD, and hippocampal expression of activity-regulated cytoskeleton-associated protein paralleled the severity of WDS. The results suggest that downstream serotonergic synaptic activation is less pronounced in GEPR-9s than in SD and that the serotonergic agent may directly activate postsynaptic 5-HT2A receptors in both strains.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R103
Ritanserin, powder
C27H25F2N3OS