EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
The kirromycin gene cluster of Streptomyces collinus Tü 365 codes for an aspartate-alpha-decarboxylase, KirD, which is involved in the biosynthesis of the precursor beta-alanine.
PMID 19609288
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K1507
Kirromycin from Streptomyces collinus
C43H60N2O12