EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of thioesters from carboxylic acids via acyloxyphosphonium intermediates with benzyltriethylammonium tetrathiomolybdate as the sulfur transfer reagent.
PMID 19618897
Abstract
An efficient protocol is reported for the synthesis of thioesters from carboxylic acids with use of acyloxy phosphonium salts as intermediates and benzyltriethylammonium tetrathiomolybdate as the sulfur transfer reagent.