EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
Production of n-octanoyl-modified ghrelin in cultured cells requires prohormone processing protease and ghrelin O-acyltransferase, as well as n-octanoic acid.
PMID 19628676
Abstract
Ghrelin was originally isolated from rat stomach as an endogenous ligand for the GH secretagogue receptor. The major active form of ghrelin is a 28-amino acid peptide modified by an n-octanoic acid on the serine 3 residue, and this lipid modification is essential for the biological activity of ghrelin. However, it is not clear whether prohormone convertase (PC) and ghrelin O-acyltransferase (GOAT) are the minimal requirements for synthesis of acyl-modified ghrelin in cultured cells. By using three cultured cell lines, TT, AtT20 and COS-7, in which the expression levels of processing proteases and GOAT vary, we examined the processing patterns of ghrelin precursor. We found that not only PC1/3 but also both PC2 and furin could process proghrelin to the 28-amino acid ghrelin. Moreover, the presence of PC and GOAT in the cells, as well as n-octanoic acid in the culture medium, was necessary to produce n-octanoyl ghrelin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O6877
Octanoyl coenzyme A lithium salt hydrate, ≥95% (HPLC)
C29H50N7O17P3S · xLi+ · yH2O