EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Cyclin-dependent kinase 5 inhibitors: inhibition of dopamine transporter activity.
PMID 19628755
Abstract
Cyclin-dependent kinase (Cdk) 5 reduces the rewarding properties of psychostimulants by dampening postsynaptic dopamine (DA) receptor signaling. Cdk5 is also present in midbrain DA neurons, where the DA transporter (DAT) is localized and limits DA neurotransmission by removing extracellular DA. Here, we tested the hypothesis that Cdk5 could also affect the disposition of DA by regulating DAT activity. Incubation of rat dorsal striatal (dSTR) synaptosomes with the Cdk5 inhibitors roscovitine, olomoucine, and 4-{[(7-oxo-6,7-dihydro-8H-[1,3]thiazolo[5,4-e]indol-8-ylidene)methyl]amino}-N-(2-pyridyl)benzenesulfonamide (GW8510) or the inactive congener iso-olomoucine resulted in a rapid, concentration-dependent inhibition of specific [3H]DA uptake. However, roscovitine was the only inhibitor that did not also decrease [3H]2beta-carbomethoxy-3beta-(4-fluorophenyl)tropane (WIN35,428) binding to dSTR DATs. Roscovitine-induced inhibition of dSTR [3H]DA uptake was explained by decreased maximal uptake velocity, without a change in cell-surface DAT levels. Roscovitine did not enhance [3H]DA release mediated by either DAT reverse-transport or Ca(2+) channels in dSTR slices. Instead, roscovitine enhanced spontaneous [3H]DA outflow and inhibited DAT-mediated [3H]DA reaccumulation into dSTR slices. To explore the involvement of Cdk5 in roscovitine-induced down-regulation of DAT activity, Cdk5 protein was knocked down via Cdk5-small interfering RNA by as much as 86% in porcine aortic endothelial cells stably expressing human (h)DATs. However, Cdk5 depletion did not alter hDAT activity. Taken together, our results suggest that roscovitine inhibits DAT activity independently of Cdk5; therefore, results obtained with such inhibitors should be interpreted with caution. Our study is the first to demonstrate that Cdk5 inhibitors reduce brain DAT activity via a mechanism that is independent of DAT trafficking and reverse-transport.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C124
β-CFT naphthalenedisulfonate monohydrate, solid
C16H20FNO2 · C10H8O6S2 · H2O