EMAIL THIS PAGE TO A FRIEND
Organic letters
An efficient and economic asymmetric synthesis of (+)-nootkatone, tetrahydronootkatone, and derivatives.
PMID 19630428
Abstract
A facile route to enantiomerically pure (+)-nootkatone and derivatives has been established through conjunctive stereoselective Grignard/anionic oxy-Cope (AOC) reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74437
(+)-Nootkatone, ≥99.0% (GC)
C15H22O
93785
(+)-Nootkatone, technical, ≥85% (GC)
C15H22O