Molecular brain
Regulation of endosomal motility and degradation by amyotrophic lateral sclerosis 2/alsin.
PMID 19630956
Dysfunction of alsin, particularly its putative Rab5 guanine-nucleotide-exchange factor activity, has been linked to one form of juvenile onset recessive familial amyotrophic lateral sclerosis (ALS2). Multiple lines of alsin knockout (ALS2(-/-)) mice have been generated to model this disease. However, it remains elusive whether the Rab5-dependent endocytosis is altered in ALS2(-/-) neurons. To directly examine the Rab5-mediated endosomal trafficking in ALS2(-/-) neurons, we introduced green fluorescent protein (GFP)-tagged Rab5 into cultured hippocampal neurons to monitor the morphology and motility of Rab5-associated early endosomes. Here we report that Rab5-mediated endocytosis was severely altered in ALS2(-/-) neurons. Excessive accumulation of Rab5-positive vesicles was observed in ALS2(-/-) neurons, which correlated with a significant reduction in endosomal motility and augmentation in endosomal conversion to lysosomes. Consequently, a significant increase in endosome/lysosome-dependent degradation of internalized glutamate receptors was observed in ALS2(-/-) neurons. These phenotypes closely resembled the endosomal trafficking abnormalities induced by a constitutively active form of Rab5 in wild-type neurons. Therefore, our findings reveal a negatively regulatory mechanism of alsin in Rab5-mediated endosomal trafficking, suggesting that enhanced endosomal degradation in ALS2(-/-) neurons may underlie the pathogenesis of motor neuron degeneration in ALS2 and related motor neuron diseases.