EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Copper(II) complexes of coumarin-derived Schiff bases and their anti-Candida activity.
PMID 19631386
Abstract
The condensation of 7-amino-4-methyl-coumarin (1) with a number of substituted salicylaldehydes yielded a series of Schiff bases (2a-2k) in good yields. Subsequent reaction of these ligands with copper(II) acetate yielded Cu(II) complexes (3a-3k) and some were characterised using X-ray crystallography. All of the free ligands and their metal complexes were tested for their anti-Candida activity. A number of the ligands and complexes exhibited anti-Candida activity comparable to that of the commercially available antifungal drugs, ketoconazole and Amphotericin B.
Related Materials