EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Parallel synthesis of chiral pentaamines and pyrrolidine containing bis-heterocyclic libraries. Multiple scaffolds with multiple building blocks: a double diversity for the identification of new antitubercular compounds.
PMID 19632841
Abstract
Combinatorial chemistry offers a unique opportunity for the synthesis and screening of large numbers of compounds and significantly enhances the prospect of finding new drugs. Collaborative efforts with the Tuberculosis Antimicrobial Acquisition & Coordinating Facility (TAACF), have led to the identification of submicromolar novel antitubercular hits. Chiral pentaamines and bis-heterocyclic compounds with 90-100% inhibition against Mycobacterium tuberculosis strain H(37)R(v) were identified. Some of the identified compounds are more active than the existing drug ethambutol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263419
3,5-Bis(trifluoromethyl)phenylacetic acid, 98%
C10H6F6O2
248045
3-Fluorophenylacetic acid, 98%
C8H7FO2