EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of 3-N-sugar-substituted-2, 4(1H,3H)-quinazolinedionesas anti-angiogenesis agents.
PMID 19633615
Abstract
A series of novel 3-N-sugar-substituted quinazolinediones were synthesizedthrough the cyclization of the intermediate 2-aminobenzamides using triphosgene as the condensing reagent. Their anti-angiogenesis activities were investigated. The compound 3-(2'-aminoglucosyl)-2,4-(1H,3H)-quinazolinedione, (5d) showed good anti-angiogenesis activity.
Related Materials