EMAIL THIS PAGE TO A FRIEND
Prescrire international
Docosanol: new drug. Herpes labialis: barely more effective than an excipient.
PMID 19637417
Abstract
(1) Herpes is a contagious, recurrent viral infection of the skin and mucous membranes. In immunocompetent patients the recurrences can be troublesome but they heal spontaneously. Management is mainly based on lifestyle measures. Local application of an antiviral drug such as aciclovir has modest effects. It reduces healing time by about 2 days provided treatment is started as soon as the first symptoms appear; (2) Docosanol, a fatty alcohol, was recently authorized in France for treatment of episodes of herpes labialis; (3) A trial in 474 patients showed no tangible difference between docosanol and 5% aciclovir in reducing healing time; (4) Clinical evaluation also includes two trials versus an excipient (polyethylene glycol) including 370 and 373 patients. The median healing time was reduced by less than a day; (5) In these trials, the adverse effects of docosanol were similar to those of the excipients. In particular, docosanol cream contains excipients that can provoke allergic reactions; (6) In practice, docosanol cream is barely or no more effective than an excipient in treating acute episodes of herpes labialis. Lifestyle measures are still the cornerstone of herpes management.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
169102
1-Docosanol, 98%
C22H46O