EMAIL THIS PAGE TO A FRIEND
Sheng wu gong cheng xue bao = Chinese journal of biotechnology
[Preparation of aminated porous resin and for bilirubin adsorption].
PMID 19637640
Abstract
Polystyrene microspheres (PS) were successfully prepared by suspension polymerization processes. Chloroacetylated polystyrene has been prepared by Friedel-Crafts acetylation of PS with chloroacetyl chloride. In this report, carcinogenic compound (chloromethylether etc.) was avoided. The effects of solvent, catalyst, acylating agent and reaction time were studied. Novel adsorption resins were obtained by synthesis of chloroacetylated polystyrene with amine. The influences of solvent, amine reagent and reaction time on ion exchange capacity were investigated. Under the optimized reaction condition, the ion exchange capacity of the prepared resins was 4.1587 mmol/g. The maximum amount of adsorbed bilirubin was 30.85 mg/g, the adsorption percentage was 80%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
22880
Chloroacetyl chloride, purum, ≥99.0% (GC)
C2H2Cl2O
104493
Chloroacetyl chloride, 98%
C2H2Cl2O