EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on chemical constituents from roots of Peucedanum praeruptorum III].
PMID 19639787
Abstract
To study the chemical constituents in roots of Peucedanum praeruptorum. The constituents were isolated by column chromatography on silica gel and ODS, and identified by spectroscopic methods. Seven compounds, alpha-D-glucopyranose-1-hexadecanoate (1), D-mannitol monohexadecanoate (2), adenosine (3), butyric acid (4), eleutheroside B, (5), apiosylskimmin (6), and mannitol (7) were isolated and identified. Compounds 1-5 were isolated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90974
Eleutheroside B, analytical standard
C17H24O9