EMAIL THIS PAGE TO A FRIEND
International Journal of Peptide and Protein Research
Synthesis of insulin-like growth factor I using N-methyl pyrrolidinone as the coupling solvent and trifluoromethane sulphonic acid cleavage from the resin.
PMID 1964154