EMAIL THIS PAGE TO A FRIEND
Journal of environmental quality
Field-scale cleanup of atrazine and cyanazine contaminated soil with a combined chemical-biological approach.
PMID 19643745
Abstract
A former agrichemical dealership in western Nebraska was suspected of having contaminated soil. Our objective was to characterize and remediate the contaminated site by a combined chemical-biological approach. This was accomplished by creating contour maps of the on-site contamination, placing the top 60 cm of contaminated soil in windrows and mixing with a mechanical high-speed mixer. Homogenized soil containing both atrazine [6-chloro-N-ethyl-N'-isopropyl-1,3,5-triazine-2,4-diamine] and cyanazine {2-[[4-chloro-6-(ethylamino)-1,3,5-triazin-2-yl] amino]-2-methylpropanenitrile} was then used in laboratory investigations to determine optimum treatments for pesticide destruction. Iron suspension experiments verified that zerovalent iron (Fe(0)) plus ferrous sulfate (FeSO(4).7H(2)O) removed more than 90% of both atrazine and cyanazine within 14 d. Liquid chromatography/mass spectrometry (LC/MS) analysis of the atrazine solution after treating with Fe(0) and ferrous sulfate identified several degradation products commonly associated with biodegradation (i.e., deethlyatrazine (DEA), deisopropylatrazine (DIA), hydroxyatrazine (HA), and ammelines). Biological treatment evaluated emulsified soybean [Glycine max (L.) Merr.] oil (EOS) as a carbon source to stimulate biodegradation in static soil microcosms. Combining emulsified soybean oil with the chemical amendments resulted in higher destruction efficiencies (80-85%) and reduced the percentage of FeSO(4) needed. This chemical-biological treatment (Fe(0) + FeSO(4) + EOS, EOS Remediation, Raleigh, NC) was then applied with water to 275 m(3) of contaminated soil in the field. Windrows were tightly covered with clear plastic to increase soil temperature and maintain soil water content. Temporal sampling (0-342 d) revealed atrazine and cyanazine concentrations decreased by 79 to 91%. These results provide evidence that a combined chemical-biological approach can be used for on-site, field-scale treatment of pesticide-contaminated soil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45407
Cyanazine, PESTANAL®, analytical standard
C9H13ClN6
442530
Cyanazine, analytical standard
C9H13ClN6