EMAIL THIS PAGE TO A FRIEND
Neurotoxicity research
Effects of omega-3 essential fatty acids (omega-3 EFAs) on motor disorders and memory dysfunction typical neuroleptic-induced: behavioral and biochemical parameter.
PMID 19644727
Abstract
The effects of fish oil supplementation on motor disorders, memory dysfunction, and lipid peroxidation (LP) induced by typical neuroleptics were studied. Wistar rats received a suspension prepared with fish oil containing omega-3 fatty acids, water, and Tween 80 (1%) in the place of drinking water (FO group) or vehicle (C group) for 8 weeks. After 4 weeks of treatment, half of the animals of both groups were treated with haloperidol (H and FO + H groups; experiment 1), fluphenazine (F and FO + F groups; experiment 2), or vehicle (C group), administered once a week (12 mg/kg/im) for 4 weeks, maintaining the treatment with FO. Extrapyramidal motor disorders by haloperidol and fluphenazine were observed by an increase in vacuous chewing movements and catalepsy (P < 0.05). These effects were reduced by FO treatment (P < 0.05). Both neuroleptics displayed impairment in memory retention observed by latency time to find the original location of platform in water-maze task, after 4 days of training performed in the last treatment week. This effect was reduced by FO (P < 0.05) to both haloperidol and fluphenazine treatments. Haloperidol increased the LP in plasma and hippocampus, and these effects were decreased by FO treatment (P < 0.05). Fluphenazine increased the LP in plasma and substantia nigra, which were completely decreased by FO treatment (P < 0.05). The FO decreased the motor disorders, memory dysfunction, and oxidative damage typical neuroleptic-induced. Our results indicate that FO exhibits a neuroprotector role useful on diseases related to oxidative damages, and may be considered in the prevention of motor and memory side effects induced by the antipsychotic treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0100100 Haloperidol decanoate, European Pharmacopoeia (EP) Reference Standard
C31H41ClFNO3