EMAIL THIS PAGE TO A FRIEND
Journal of clinical pharmacy and therapeutics
Pharmacokinetic profile of talipexole in healthy volunteers is not altered when it is co-administered with Madopar (co-beneldopa).
PMID 19646081
Abstract
To investigate the pharmacokinetics of talipexole hydrochloride tablets and the potential influence of Madopar (benserazide and levodopa combination; co-beneldopa) tablets on talipexole's pharmacokinetics when the two tablets are co-administered orally to healthy Chinese volunteers. A sensitive liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed to measure talipexole concentration in human plasma in an open-label, randomized, two-way crossover, single-dose study, with 1-week washout period. Healthy Chinese volunteers were randomized to receive talipexole tablets either alone or together with Madopar tablets by oral administration after an overnight fast. Serial blood samples were collected for a period of 36 h after the administration. Pharmacokinetic parameters C(max), t(max), t(1/2z), mean residence time (MRT), AUC(0-tau), AUC(0-infinity), CL(z)/F and V(z)/F were determined under the non-compartmental model. Pharmacokinetic values of talipexole administered alone to the subjects were compared with those administered simultaneously with Madopar to determine whether or not the differences were statistically significant. The subjects experienced mild gastrointestinal irritation when talipexole was administered alone as well as together with Madopar. For talipexole hydrochloride, there were no significant differences in the pharmacokinetic values between the two administrations. No pharmacokinetic differences based on gender were observed either. A single oral dose of Madopar co-administered with talipexole does not significantly change talipexole's pharmacokinetics in human.