EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Effects of inhibition of neuronal nitric oxide synthase on basal retinal blood flow regulation.
PMID 19646435
Abstract
Nitric oxide (NO) has been observed to regulate blood flow under basal and stimulated conditions in the retina. Recent evidence suggests that NO produced by neuronal nitric oxide synthase (nNOS) may regulate blood flow in addition to that produced by endothelial nitric oxide synthase (eNOS). The objective of the current study was to investigate the contribution of NO produced by nNOS in the regulation of basal retinal blood flow. A non-specific NOS inhibitor N (G)-nitro-l-arginine methyl ester (l-NAME) and the specific nNOS inhibitors 1-(2-trifluoromethylphenyl) imidazole (TRIM) and (4S)-N-(4-amino-5 [aminoethyl] aminopentyl)-N-nitroguanidine (AAAN) were injected into the vitreous (intravitreal) of Long-Evans rats. Vessel diameters, velocities and volumetric blood flow rates (VBF) in the retinal circulation were determined prior to and in 30-min intervals for 4-4.5h after injection. In addition, the basal amount of nNOS in the rat retina was quantified using a specific enzyme linked immunoassay (ELISA). Treatment with l-NAME and TRIM significantly decreased diameters and VBF. Compared with saline, treatment with l-NAME and TRIM produced a significant (p<0.001) decrease of approximately 12-17% in vessel diameters. Treatment with AAAN significantly decreased vessel diameters and venous VBF. Compared with saline AAAN produced a significant decrease of approximately 7% in arterial (p<0.001) and 5% in venous (p=0.011) diameters, respectively. The amount of nNOS in the rat retina was 0.17+/- 0.0147 pmol mg(-1) of dry retina. The results suggest that though inhibition of nNOS decreases basal diameters, constant VBF is maintained in the retinal circulation.