EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Scope and mechanism of intramolecular aziridination of cyclopent-3-enyl-methylamines to 1-azatricyclo[2.2.1.0(2,6)]heptanes with lead tetraacetate.
PMID 19653649
Abstract
A series of seven cyclopent-3-en-1-ylmethylamines bearing one, two, or three methyl substituents at the C2, C3, C4, or C(alpha) positions, including the unsubstituted parent, was accessed by ring-closing metatheses of alpha,alpha-diallylacetonitrile (or methallyl variants) and alpha,alpha-diallylacetone followed by hydride reductions or reductive amination, or by Curtius degradations of alpha,alpha-dimethyl- and 2,2,3-trimethylcyclopent-3-enylacetic acids. Oxidation of the primary amines with Pb(OAc)(4) in CH(2)Cl(2), CHCl(3) or benzene in the presence of K(2)CO(3) effected efficient intramolecular aziridinations, in all cases except the alpha-methyl analogue (16), to form the corresponding 1-azatricyclo[2.2.1.0(2,6)]heptanes, including the novel monoterpene analogues, 1-azatricyclene and the 2-azatricyclene enantiomers. The cumulative rate increases of aziridination reactions observed by (1)H NMR spectroscopy in CDCl(3) resulting from the presence of one or two methyl groups on the cyclopentene double bond, in comparison to the rate of the unsubstituted parent amine (1:17.5:>280), indicate a highly electrophilic intermediate as the nitrene donor and a symmetrical aziridine-like transition state. A mechanism is outlined in which the amine displaces an acetate ligand from Pb(OAc)(4) to form a lead(IV) amide intermediate RNHPb(OAc)(3) proposed as the actual aziridinating species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185191
Lead(IV) acetate, reagent grade, 95%
C8H12O8Pb