PloS one
Apoptosis of CD4+ CD25(high) T cells in type 1 diabetes may be partially mediated by IL-2 deprivation.
PMID 19654878
Abstract
Type 1 diabetes (T1D) is a T-cell mediated autoimmune disease targeting the insulin-producing pancreatic beta cells. Naturally occurring FOXP3(+)CD4(+)CD25(high) regulatory T cells (T(regs)) play an important role in dominant tolerance, suppressing autoreactive CD4(+) effector T cell activity. Previously, in both recent-onset T1D patients and beta cell antibody-positive at-risk individuals, we observed increased apoptosis and decreased function of polyclonal T(regs) in the periphery. Our objective here was to elucidate the genes and signaling pathways triggering apoptosis in T(regs) from T1D subjects. Gene expression profiles of unstimulated T(regs) from recent-onset T1D (n = 12) and healthy control subjects (n = 15) were generated. Statistical analysis was performed using a Bayesian approach that is highly efficient in determining differentially expressed genes with low number of replicate samples in each of the two phenotypic groups. Microarray analysis showed that several cytokine/chemokine receptor genes, HLA genes, GIMAP family genes and cell adhesion genes were downregulated in T(regs) from T1D subjects, relative to control subjects. Several downstream target genes of the AKT and p53 pathways were also upregulated in T1D subjects, relative to controls. Further, expression signatures and increased apoptosis in T(regs) from T1D subjects partially mirrored the response of healthy T(regs) under conditions of IL-2 deprivation. CD4(+) effector T-cells from T1D subjects showed a marked reduction in IL-2 secretion. This could indicate that prior to and during the onset of disease, T(regs) in T1D may be caught up in a relatively deficient cytokine milieu. In summary, expression signatures in T(regs) from T1D subjects reflect a cellular response that leads to increased sensitivity to apoptosis, partially due to cytokine deprivation. Further characterization of these signaling cascades should enable the detection of genes that can be targeted for restoring T(reg) function in subjects predisposed to T1D.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54675 Stator Face Assembly, for use with (7125)