EMAIL THIS PAGE TO A FRIEND
The American journal of Chinese medicine
Inhibition of NMDA receptors underlies the neuroprotective effect of ginsenoside Rb3.
PMID 19655413
Abstract
In order to investigate the mechanisms underlying the neuroprotective effect of ginsenoside Rb3, rat hippocampal neurons were primarily cultured, and exposed to 1 mM N-methyl-D-aspartate (NMDA), cell viability and lactate dehydrogenase leakage were measured. Ca2+ influx was determined by calcium imaging with a laser confocal microscopy. The influences of ginsenoside Rb3 on these variables were examined. Patch-clamp technique was used to observe the effects of ginsenoside Rb3 on NMDA-evoked current. The results show that treatment of Rb3 raised the neuronal viability, reduced the leakage of lactate dehydrogenase, and inhibited NMDA-elicited Ca2+ influx in a dose-dependent manner. In the presence of Rb3, NMDA-evoked peak current was inhibited, and Ca2+-induced desensitization of NMDA current was facilitated. It is suggested that ginsenoside Rb3 could exert a neuroprotective role on hippocampal neurons, a role which was partly mediated by the facilitation of Ca2+-dependent deactivation of NMDA receptors, and the resultant reduction of intracellular free Ca2+ level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42635
Ginsenoside Rb3, analytical standard
C53H90O22