EMAIL THIS PAGE TO A FRIEND
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc
Evidence for a porcine respiratory coronavirus, antigenically similar to transmissible gastroenteritis virus, in the United States.
PMID 1965638
Abstract
A respiratory variant of transmissible gastroenteritis virus (TGEV), designated PRCV-Ind/89, was isolated from a swine breeding stock herd in Indiana. The virus was readily isolated from nasal swabs of pigs of different ages and induced cytopathology on primary porcine kidney cells and and on a swine testicular (ST) cell line. An 8-week-old pig infected oral/nasally with the respiratory variant and a contact pig showed no signs of respiratory or enteric disease. These pigs did not shed virus in feces but did shed the agent from the upper respiratory tract for approximately 2 weeks. Baby pigs from 2 separate litters (2 and 3 days old) also showed no clinical signs following oral/nasal inoculation with PRCV-Ind/89. In a third litter, 5 of 7 piglets (5 days old) infected either oral/nasally or by stomach tube developed a transient mild diarrhea with villous atrophy. However, virus was not isolated from rectal swabs or ileal homogenates of these piglets, and viral antigen was not detected in the ileum by fluorescent antibody staining even though the virus was easily recovered from nasal swabs and lung tissue homogenates. Swine antisera produced against PRCV-Ind/89 or enteric TGEV cross-neutralized either virus. In addition, an anti-peplomer monoclonal antibody, 4F6, that neutralizes TGEV also neutralized the PRCV-Ind/89 isolate. Radioimmunoassays with a panel of monoclonal antibodies indicated that the Indiana respiratory variant and the European PRCV are antigenically similar.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
ST, 92040221