EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Of two make one: the biosynthesis of phenazines.
PMID 19658148
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1761
Chorismic acid from Enterobacter aerogenes, ≥80%
C10H10O6