EMAIL THIS PAGE TO A FRIEND
Neurobiology of Disease
Biochemical and immunohistochemical analysis of an Alzheimer's disease mouse model reveals the presence of multiple cerebral Abeta assembly forms throughout life.
PMID 19660551