EMAIL THIS PAGE TO A FRIEND
Journal of biomolecular structure & dynamics
Accessibility of tryptophan residues in immunoglobulin M as an index of its conformational changeability.
PMID 1966064
Abstract
Demonstrated herein is the possibility of using the accessibility of tryptophan (Trp) residues in immunoglobulin M (IgM) upon modification with Koshland reagent (2-hydroxy-5-nitrobenzyl bromide) as an index of the conformational changeability of IgM. Of fourteen Trp's in the native IgM (per HL-region) only one appeared to be most accessible, evidently Trp312 in the mu-chain. Irreversible acidic and thermal conformational transitions in IgM increase the number of accessible Trp's approximately two-fold. Following partial enzymatic deglycosylation of IgM, deep scission of mannose in particular, all Trp's become inaccessible. Modification of the most accessible Trp increases 2-3 fold the number of tyrosine residues readily accessible upon nitration with tetranitromethane. Modification of four trp's using spin-label method data causes a sharp reduction of the mobility of the C mu 3 domain and a simultaneous decrease in the solubility of modified IgM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
162906
2-Hydroxy-5-nitrobenzyl bromide, 90%
C7H6BrNO3